ENA514506731 Enamine

7-fluoronaphthalen-1-ol

  • MDL number MFCD00077540
  • Cas Number : 3132-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-140
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-302206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-302206
Availability details for
Quantity
  • Availability
  • Add to Cart