ENA514506789 Enamine

(6-aminohexyl)(methyl)amine

  • MDL number MFCD12796825
  • Cas Number : 2997-06-0
  • Molecular Weight : 130.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-304314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-304314
Availability details for
Quantity
  • Availability
  • Add to Cart