ENA514506975 Enamine

2-(ethylamino)propanenitrile hydrochloride

  • MDL number MFCD30342611
  • Cas Number : 1989659-41-7
  • Molecular Weight : 134.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-305148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-305148
Availability details for
Quantity
  • Availability
  • Add to Cart