ENA514507058 Enamine

5-[(1E)-3-methoxy-3-oxoprop-1-en-1-yl]furan-2-carboxylic acid

  • Cas Number : 125812-00-2
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-305412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-305412
Availability details for
Quantity
  • Availability
  • Add to Cart