ENA514507061 Enamine

tert-butyl 2-[(2,2-dimethylpropyl)amino]acetate

  • MDL number MFCD14593233
  • Cas Number : 765957-52-6
  • Molecular Weight : 201.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-305418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-305418
Availability details for
Quantity
  • Availability
  • Add to Cart