ENA514507082 Enamine

2-amino-2-(2,3-dichlorophenyl)acetic acid

  • MDL number MFCD02662393
  • Cas Number : 318270-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-305456

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-305456
Availability details for
Quantity
  • Availability
  • Add to Cart