ENA514507142 Enamine

2-{4-[(tert-butoxy)carbonyl]-1,1-dioxo-1lambda6-thiomorpholin-3-yl}acetic acid

  • MDL number MFCD22396032
  • Cas Number : 1542480-23-8
  • Molecular Weight : 293.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106970
Availability details for
Quantity
  • Availability
  • Add to Cart