ENA514507156 Enamine

methyl 3-{[(benzyloxy)carbonyl]amino}thiophene-2-carboxylate

  • MDL number MFCD06223263
  • Cas Number : 778602-61-2
  • Molecular Weight : 291.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-305683

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-305683
Availability details for
Quantity
  • Availability
  • Add to Cart