ENA514507174 Enamine

(1S)-1-(1-benzofuran-3-yl)ethan-1-ol

  • MDL number MFCD15146065
  • Cas Number : 343614-13-1
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-305724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-305724
Availability details for
Quantity
  • Availability
  • Add to Cart