ENA514507536 Enamine

(3,4-dimethylpyrrolidin-3-yl)methanol hydrochloride

  • MDL number MFCD30342947
  • Cas Number : 1989672-09-4
  • Molecular Weight : 165.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-306833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-306833
Availability details for
Quantity
  • Availability
  • Add to Cart