ENA514507754 Enamine

[(1R,5S,6R)-3-oxabicyclo[3.1.0]hexan-6-yl]methanol

  • MDL number MFCD27986972
  • Cas Number : 135577-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-307189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-307189
Availability details for
Quantity
  • Availability
  • Add to Cart