ENA514507762 Enamine

4-fluoro-3-iodopyridin-2-amine

  • MDL number MFCD28368511
  • Cas Number : 1805602-68-9
  • Molecular Weight : 238

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-306374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-306374
Availability details for
Quantity
  • Availability
  • Add to Cart