ENA517161259 Enamine

2,2,2-trifluoroethyl N-[(2,2,2-trifluoroethoxy)carbonyl]carbamate

  • MDL number MFCD30345023
  • Cas Number : 2031269-45-9
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-306466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-306466
Availability details for
Quantity
  • Availability
  • Add to Cart