ENA517161415 Enamine

3-bromo-4,5-dicyclopropyl-4H-1,2,4-triazole

  • MDL number MFCD30344967
  • Cas Number : 2031261-09-1
  • Molecular Weight : 228.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-309113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-309113
Availability details for
Quantity
  • Availability
  • Add to Cart