ENA517161441 Enamine

1,4,8-oxadiazecan-5-one

  • MDL number MFCD30345180
  • Cas Number : 2044713-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-310026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-310026
Availability details for
Quantity
  • Availability
  • Add to Cart