ENA517161457 Enamine

tert-butyl N-methyl-N-(2,2,2-trifluoroethoxy)carbamate

  • MDL number MFCD30345202
  • Cas Number : 2031269-17-5
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-310251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-310251
Availability details for
Quantity
  • Availability
  • Add to Cart