ENA517161613 Enamine

ethyl 2-tert-butylcyclopropane-1-carboxylate

  • MDL number MFCD30345020
  • Cas Number : 21692-49-9
  • Molecular Weight : 170.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-306440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-306440
Availability details for
Quantity
  • Availability
  • Add to Cart