ENA517161648 Enamine

5-methyl-4-nitrothiophene-2-carboxamide

  • MDL number MFCD01025640
  • Cas Number : 36050-10-9
  • Molecular Weight : 186.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-307088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-307088
Availability details for
Quantity
  • Availability
  • Add to Cart