ENA517161858 Enamine

4-(propan-2-yl)-1H-imidazole-2-thiol

  • MDL number MFCD19236884
  • Cas Number : 56659-00-8
  • Molecular Weight : 142.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-102522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102522
Availability details for
Quantity
  • Availability
  • Add to Cart