ENA517161860 Enamine

2-(2-methylcyclopropyl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD21198428
  • Cas Number : 1496334-19-0
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-257050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-257050
Availability details for
Quantity
  • Availability
  • Add to Cart