ENA517161924 Enamine

2,4-dimethyl-5,6,7,8-tetrahydroquinolin-5-one

  • MDL number MFCD13189692
  • Cas Number : 34192-19-3
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268133

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268133
Availability details for
Quantity
  • Availability
  • Add to Cart