ENA517162020 Enamine

thieno[2,3-c]pyridine-4-carboxylic acid

  • MDL number MFCD27921580
  • Cas Number : 1554074-84-8
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-309564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-309564
Availability details for
Quantity
  • Availability
  • Add to Cart