ENA517162088 Enamine

1-acetyl-1,2,3,4-tetrahydroquinolin-4-one

  • MDL number MFCD20318871
  • Cas Number : 64142-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113015
Availability details for
Quantity
  • Availability
  • Add to Cart