ENA517162431 Enamine

4-chloro-3-methylquinolin-5-amine

  • MDL number MFCD30345255
  • Cas Number : 2031268-87-6
  • Molecular Weight : 192.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-310739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-310739
Availability details for
Quantity
  • Availability
  • Add to Cart