ENA517162787 Enamine

methyl 3-methyl-5-oxohexanoate

  • MDL number MFCD24689265
  • Cas Number : 14983-18-7
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-312300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-312300
Availability details for
Quantity
  • Availability
  • Add to Cart