ENA517162827 Enamine

methyl 4-{[(tert-butoxy)carbonyl]amino}cyclopent-1-ene-1-carboxylate

  • MDL number MFCD24532039
  • Cas Number : 1169936-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-312466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-312466
Availability details for
Quantity
  • Availability
  • Add to Cart