ENA517162914 Enamine

5-fluoro-1-(methoxymethyl)-1H-indole

  • MDL number MFCD21195622
  • Cas Number : 1488594-93-9
  • Molecular Weight : 179.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-312746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-312746
Availability details for
Quantity
  • Availability
  • Add to Cart