ENA517163090 Enamine

1-sulfanylpentan-3-one

  • MDL number MFCD24693518
  • Cas Number : 123728-54-1
  • Molecular Weight : 118.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-311598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-311598
Availability details for
Quantity
  • Availability
  • Add to Cart