ENA517163102 Enamine

{[1,3]thiazolo[5,4-b]pyridin-2-yl}hydrazine

  • MDL number MFCD00174244
  • Cas Number : 57135-11-2
  • Molecular Weight : 166.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-311623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-311623
Availability details for
Quantity
  • Availability
  • Add to Cart