ENA517163143 Enamine

2-(cyclopropylmethoxy)-3,3,3-trifluoropropan-1-amine

  • MDL number MFCD12047281
  • Cas Number : 1157932-10-9
  • Molecular Weight : 183.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68581

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68581
Availability details for
Quantity
  • Availability
  • Add to Cart