ENA580348335 Enamine

2-fluoro-3-methylpyridine-4-carbaldehyde

  • MDL number MFCD16611269
  • Cas Number : 1211581-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195036
Availability details for
Quantity
  • Availability
  • Add to Cart