ENA580348364 Enamine

1-(4-methoxypyridin-2-yl)ethan-1-one

  • MDL number MFCD10697664
  • Cas Number : 59576-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211520
Availability details for
Quantity
  • Availability
  • Add to Cart