ENA580348411 Enamine

4-(aminomethyl)-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD30476064
  • Cas Number : 1936337-16-4
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-227420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-227420
Availability details for
Quantity
  • Availability
  • Add to Cart