ENA580349410 Enamine

methyl 4-aminocyclopent-1-ene-1-carboxylate hydrochloride

  • MDL number MFCD30476263
  • Cas Number : 99237-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-313792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-313792
Availability details for
Quantity
  • Availability
  • Add to Cart