ENA580350085 Enamine

(2S,4R)-4-hydroxypyrrolidine-2-carboxamide hydrochloride

  • MDL number MFCD26404526
  • Cas Number : 32934-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-314410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-314410
Availability details for
Quantity
  • Availability
  • Add to Cart