ENA580350295 Enamine

tert-butyl N-(2-bromo-4-methyl-1,3-thiazol-5-yl)carbamate

  • MDL number MFCD24387253
  • Cas Number : 1255095-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-313932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-313932
Availability details for
Quantity
  • Availability
  • Add to Cart