ENA580353268 Enamine

7-bromoisoquinoline-1-carbonitrile

  • MDL number MFCD18253941
  • Cas Number : 1204334-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-320468

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-320468
Availability details for
Quantity
  • Availability
  • Add to Cart