ENA580354025 Enamine

4-bromo-5-fluoro-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD30487861
  • Cas Number : 2044902-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-321679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-321679
Availability details for
Quantity
  • Availability
  • Add to Cart