ENA580354238 Enamine

tert-butyl N-{[(1s,3s)-3-hydroxycyclobutyl]methyl}carbamate

  • MDL number MFCD16660648
  • Cas Number : 917827-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-321463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-321463
Availability details for
Quantity
  • Availability
  • Add to Cart