ENA580354468 Enamine

1-(4-bromophenyl)-5-phenyl-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD13378722
  • Cas Number : 92425-18-8
  • Molecular Weight : 344.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-322593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322593
Availability details for
Quantity
  • Availability
  • Add to Cart