ENA580354604 Enamine

ethyl 1-bromoimidazo[1,5-a]pyridine-3-carboxylate

  • MDL number MFCD06739184
  • Cas Number : 885276-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-323129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-323129
Availability details for
Quantity
  • Availability
  • Add to Cart