ENA580357868 Enamine

2-(oxan-4-yloxy)ethane-1-sulfonyl chloride

  • MDL number MFCD19676184
  • Cas Number : 1495584-23-0
  • Molecular Weight : 228.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-330399

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-330399
Availability details for
Quantity
  • Availability
  • Add to Cart