ENA580357985 Enamine

4-(4-bromophenyl)oxane

  • MDL number MFCD12913643
  • Cas Number : 811828-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-330516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-330516
Availability details for
Quantity
  • Availability
  • Add to Cart