ENA580357998 Enamine

4-(oxan-4-yl)aniline

  • MDL number MFCD11506022
  • Cas Number : 62071-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-330529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-330529
Availability details for
Quantity
  • Availability
  • Add to Cart