ENA580359395 Enamine

4-methyl-1,2-thiazole

  • MDL number MFCD11501471
  • Cas Number : 693-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82994

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P271,P280,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
hcodes F,Xi
MSDS https://www.enaminestore.com/catalog/EN300-82994
pictograms GHS02,GHS07
hstatements H225,H303,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart