ENA638586006 Enamine

tert-butyl N-(3-ethoxy-2-oxopropyl)carbamate

  • MDL number MFCD30497476
  • Cas Number : 2007599-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-266908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266908
Availability details for
Quantity
  • Availability
  • Add to Cart