ENA638586011 Enamine

(1-cycloheptylazetidin-2-yl)methanol

  • MDL number MFCD30535591
  • Cas Number : 2060025-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-270008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-270008
Availability details for
Quantity
  • Availability
  • Add to Cart