ENA638586019 Enamine

tert-butyl 3-methyl-[1,2,4]triazolo[4,3-a]pyrazine-6-carboxylate

  • MDL number MFCD30497501
  • Cas Number : 2059999-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-303303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-303303
Availability details for
Quantity
  • Availability
  • Add to Cart