ENA638586031 Enamine

4-methyl-1-(pyridin-3-yl)pent-3-en-1-ol

  • MDL number MFCD30497515
  • Cas Number : 2060035-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-306025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-306025
Availability details for
Quantity
  • Availability
  • Add to Cart