ENA638586032 Enamine

N-(2,2-diethoxyethyl)-4-iodo-1H-pyrrole-2-carboxamide

  • MDL number MFCD30497516
  • Cas Number : 2060063-38-7
  • Molecular Weight : 352.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-306250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-306250
Availability details for
Quantity
  • Availability
  • Add to Cart